Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lim-4
Entrez Gene ID
180672
Gene Name
protein-coding
Gene Aliases
CELE_ZC64.4
Chromosome Location
Chr.X: 3869404 - 3873545 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076268.2 NP_508669.1 3 - 4 702 131
GenBank mRNA U72348.1 - 3 - 4 453 131

View Full Product Details