Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
erd-2.1
Entrez Gene ID
181228
Gene Name
ER lumen protein-retaining receptor erd-2.1
Gene Aliases
CELE_F09B9.3
Chromosome Location
Chr.X: 10156858 - 10159741 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001381076.2 NP_001366716.1 2 - 3 1023 99

View Full Product Details