Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cyp-14A4
Entrez Gene ID
187573
Gene Name
CYtochrome P450 family
Gene Aliases
CELE_R04D3.1
Chromosome Location
Chr.X: 13282043 - 13284204 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077806.4 NP_510207.1 4 - 5 876 133

View Full Product Details