Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C33G3.4
Entrez Gene ID
181517
Gene Name
Probable beta-mannosidase
Gene Aliases
CELE_C33G3.4
Chromosome Location
Chr.X: 14071162 - 14076298 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077941.7 NP_510342.1 4 - 5 598 77

View Full Product Details