Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fbxa-81
Entrez Gene ID
188841
Gene Name
F-box domain-containing protein
Gene Aliases
CELE_T24C2.4
Chromosome Location
Chr.X: 14540660 - 14542092 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078018.2 NP_510419.1 3 - 4 796 133

View Full Product Details