Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
srbc-1
Entrez Gene ID
185314
Gene Name
protein-coding
Gene Aliases
CELE_F35F10.9
Chromosome Location
Chr.V: 3298885 - 3301998 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071512.1 NP_503913.1 2 - 3 344 117

View Full Product Details