Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
stdh-1
Entrez Gene ID
182291
Gene Name
Putative steroid dehydrogenase 1
Gene Aliases
CELE_C06B3.4
Chromosome Location
Chr.V: 13906262 - 13908053 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074048.3 NP_506449.1 2 - 3 191 142

View Full Product Details