Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
clec-49
Entrez Gene ID
180298
Gene Name
C-type LECtin
Gene Aliases
CELE_W04E12.6
Chromosome Location
Chr.V: 19748961 - 19751226 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_075428.5 NP_507829.1 2 - 3 533 102

View Full Product Details