Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
egg-5
Entrez Gene ID
172007
Gene Name
Inactive protein-tyrosine phosphatase egg-5
Gene Aliases
CELE_R12E2.10
Chromosome Location
Chr.I: 4168984 - 4171589 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058915.9 NP_491316.1 4 - 5 2176 76

View Full Product Details