Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
klp-15
Entrez Gene ID
172213
Gene Name
Kinesin motor domain-containing protein
Gene Aliases
CELE_M01E11.6
Chromosome Location
Chr.I: 5577070 - 5579457 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059232.4 NP_491633.1 2 - 3 1225 157

View Full Product Details