Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mig-14
Entrez Gene ID
175020
Gene Name
Protein wntless homolog
Gene Aliases
CELE_R06B9.6
Chromosome Location
Chr.II: 13741738 - 13749983 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027104.4 NP_001022275.1 6 - 7 876 67

View Full Product Details