Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DSG1
Entrez Gene ID
403401
Gene Name
desmoglein 1
Gene Aliases
DSG-1
Chromosome Location
Chr.7: 58159283 - 58239018 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002939.1 NP_001002939.1 6 - 7 694 82
XM_005622216.3 XP_005622273.1 7 - 8 1177 82
XM_005622217.3 XP_005622274.1 7 - 8 1115 82
GenBank mRNA AF005360.1 - 6 - 7 694 82

View Full Product Details