Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SFRP2
Entrez Gene ID
475471
Gene Name
secreted frizzled related protein 2
Gene Aliases
-
Chromosome Location
Chr.15: 51541197 - 51548765 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AJ560716.1 - 3 - 3 1533 125

View Full Product Details