Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CD3E
Entrez Gene ID
442981
Gene Name
CD3e molecule
Gene Aliases
-
Chromosome Location
Chr.5: 15403724 - 15415259 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003379.1 NP_001003379.1 - 578 105
GenBank mRNA M55410.1 - - 578 105

View Full Product Details