Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ELOB
Entrez Gene ID
479873
Gene Name
elongin B
Gene Aliases
TCEB2
Chromosome Location
Chr.6: 38311082 - 38314502 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_014114679.1 XP_013970154.1 1 - 2 116 81

View Full Product Details