Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NDUFA7
Entrez Gene ID
476722
Gene Name
NADH:ubiquinone oxidoreductase subunit A7
Gene Aliases
-
Chromosome Location
Chr.20: 52892784 - 52902007 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_533926.6 XP_533926.1 1 - 2 150 60

View Full Product Details