Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HSPE1
Entrez Gene ID
478855
Gene Name
heat shock 10kDa protein 1 (chaperonin 10)
Gene Aliases
MOB4, MOBKL3, PREI3
Chromosome Location
Chr.37: 7020507 - 7063678 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_014111037.2 XP_013966512.1 7 - 8 672 84
XM_536018.6 XP_536018.2 6 - 7 534 84
XM_852308.5 XP_857401.2 5 - 6 477 84

View Full Product Details