Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BLVRB
Entrez Gene ID
476456
Gene Name
biliverdin reductase B
Gene Aliases
-
Chromosome Location
Chr.1: 113244484 - 113258170 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_533666.6 XP_533666.2 3 - 4 444 91

View Full Product Details