Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SOX30
Entrez Gene ID
479316
Gene Name
SRY-box transcription factor 30
Gene Aliases
-
Chromosome Location
Chr.4: 52587163 - 52615945 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005619250.3 XP_005619307.1 5 - 6 2195 59
XM_536454.6 XP_536454.2 4 - 5 2147 59

View Full Product Details