Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CDKN2AIPNL
Entrez Gene ID
481507
Gene Name
CDKN2A interacting protein N-terminal like
Gene Aliases
-
Chromosome Location
Chr.11: 22548186 - 22556261 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_538628.5 XP_538628.2 2 - 3 392 91

View Full Product Details