Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EIF3J
Entrez Gene ID
487532
Gene Name
eukaryotic translation initiation factor 3 subunit J
Gene Aliases
EIF3S1
Chromosome Location
Chr.30: 11207620 - 11233106 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_544656.6 XP_544656.2 2 - 3 261 135

View Full Product Details