Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RNASEK
Entrez Gene ID
607349
Gene Name
ribonuclease K
Gene Aliases
-
Chromosome Location
Chr.5: 32040334 - 32042291 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005619936.3 XP_005619993.3 2 - 3 530 73

View Full Product Details