Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC25A37
Entrez Gene ID
486115
Gene Name
solute carrier family 25 member 37
Gene Aliases
-
Chromosome Location
Chr.25: 33923108 - 33965577 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022410097.1 XP_022265805.1 4 - 5 583 65
XM_022410098.1 XP_022265806.1 4 - 5 580 65
XM_022410099.1 XP_022265807.1 3 - 4 486 65
XM_844354.5 XP_849447.1 2 - 3 630 65

View Full Product Details