Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CTSG
Entrez Gene ID
106559147
Gene Name
cathepsin G
Gene Aliases
-
Chromosome Location
Chr.8: 4483327 - 4485933 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_014115810.2 XP_013971285.1 4 - 5 660 66

View Full Product Details