Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- MGAT1
- Entrez Gene ID
-
491674
- Gene Name
- alpha-1,3-mannosyl-glycoprotein 2-beta-N-acetylglucosaminyltransferase
- Gene Aliases
-
-
- Chromosome Location
-
Chr.11: 114587 - 129014 on Build CanFam3.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_850565.4 |
XP_855658.1 |
- |
369 |
104 |
View Full Product Details