Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IGBP1
Entrez Gene ID
480950
Gene Name
immunoglobulin binding protein 1
Gene Aliases
-
Chromosome Location
Chr.X: 54558515 - 54612029 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005641443.3 XP_005641500.2 5 - 6 953 74
XM_022415678.1 XP_022271386.1 4 - 5 661 74

View Full Product Details