Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HYAL3
Entrez Gene ID
608620
Gene Name
hyaluronidase 3
Gene Aliases
-
Chromosome Location
Chr.20: 39041940 - 39047388 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005633189.3 XP_005633246.2 2 - 3 1046 117

View Full Product Details