Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- EIF3J
- Entrez Gene ID
-
487532
- Gene Name
- eukaryotic translation initiation factor 3 subunit J
- Gene Aliases
-
EIF3S1
- Chromosome Location
-
Chr.30: 11207620 - 11233106 on Build CanFam3.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_544656.6 |
XP_544656.2 |
2 - 3 |
261 |
87 |
View Full Product Details