Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLEKHO2
Entrez Gene ID
487597
Gene Name
pleckstrin homology domain containing O2
Gene Aliases
PLEKHQ1
Chromosome Location
Chr.30: 29242306 - 29264632 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_544722.6 XP_544722.2 4 - 5 522 104

View Full Product Details