Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PLAGL1
Entrez Gene ID
484018
Gene Name
PLAG1 like zinc finger 1
Gene Aliases
-
Chromosome Location
Chr.1: 35418470 - 35529230 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005615474.3 XP_005615531.1 - 1418 90
XM_005615479.3 XP_005615536.1 - 1249 90
XM_014112545.2 XP_013968020.1 - 1753 90
XM_014112555.2 XP_013968030.1 - 1791 90
XM_014112558.2 XP_013968033.1 - 1823 90
XM_014112566.2 XP_013968041.1 - 1785 90
XM_014112572.2 XP_013968047.1 - 1767 90
XM_014112577.2 XP_013968052.1 - 1679 90
XM_022417720.1 XP_022273428.1 - 1719 90
XM_022417724.1 XP_022273432.1 - 1694 90
XM_022417725.1 XP_022273433.1 - 1617 90
XM_022417731.1 XP_022273439.1 - 1751 90
XM_022417736.1 XP_022273444.1 - 1733 90
XM_022417744.1 XP_022273452.1 - 1609 90
XM_022417747.1 XP_022273455.1 - 1723 90
XM_022417748.1 XP_022273456.1 - 1532 90

View Full Product Details