Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HMGB2
Entrez Gene ID
486068
Gene Name
high mobility group box 2
Gene Aliases
-
Chromosome Location
Chr.25: 23939024 - 23941720 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_543194.4 XP_543194.2 2 - 3 388 152

View Full Product Details