Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GAB2
Entrez Gene ID
485167
Gene Name
GRB2 associated binding protein 2
Gene Aliases
-
Chromosome Location
Chr.21: 20414903 - 20605807 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_014122569.2 XP_013978044.1 9 - 10 1970 79

View Full Product Details