Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HRK
Entrez Gene ID
612984
Gene Name
harakiri, BCL2 interacting protein
Gene Aliases
-
Chromosome Location
Chr.26: 13481095 - 13501276 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022410619.1 XP_022266327.1 1 - 1 287 78
XM_848964.5 XP_854057.2 1 - 1 379 78

View Full Product Details