Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GALR2
Entrez Gene ID
483325
Gene Name
galanin receptor 2
Gene Aliases
-
Chromosome Location
Chr.9: 4559710 - 4562474 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_540444.5 XP_540444.4 - 623 85

View Full Product Details