Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AAR2
Entrez Gene ID
477218
Gene Name
AAR2 splicing factor
Gene Aliases
C24H20orf4
Chromosome Location
Chr.24: 25053200 - 25070646 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005635002.3 XP_005635059.1 3 - 4 1611 131
XM_022409009.1 XP_022264717.1 4 - 5 1272 131
XM_022409010.1 XP_022264718.1 4 - 5 1787 131
XM_022409011.1 XP_022264719.1 4 - 5 1786 131
XM_022409012.1 XP_022264720.1 4 - 5 1892 131
XM_534409.6 XP_534409.1 3 - 4 1161 131

View Full Product Details