Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MTPN
Entrez Gene ID
403487
Gene Name
myotrophin
Gene Aliases
-
Chromosome Location
Chr.16: 12316487 - 12369148 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002989.1 NP_001002989.1 - 79 144
GenBank mRNA AY266681.1 - - 79 144

View Full Product Details