Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EGR1
Entrez Gene ID
481528
Gene Name
early growth response 1
Gene Aliases
-
Chromosome Location
Chr.11: 26046195 - 26049806 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_846145.4 XP_851238.1 1 - 2 334 154

View Full Product Details