Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CNTNAP5
Entrez Gene ID
483874
Gene Name
contactin associated protein family member 5
Gene Aliases
caspr5
Chromosome Location
Chr.19: 25603433 - 26397833 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001048108.1 NP_001041573.1 1 - 2 76 61
XM_005631764.3 XP_005631821.1 1 - 2 212 61

View Full Product Details