Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GPBAR1
Entrez Gene ID
488521
Gene Name
G protein-coupled bile acid receptor 1
Gene Aliases
-
Chromosome Location
Chr.37: 24937955 - 24943546 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005640641.3 XP_005640698.2 2 - 2 3212 102

View Full Product Details