Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
B3gat3
Entrez Gene ID
100689419
Gene Name
beta-1,3-glucuronyltransferase 3 (glucuronosyltransferase I)
Gene Aliases
GlcUAT-I, Glcat-i, I79_016742
Chromosome Location
Chr.Un|NW_003614577: 235477 - 238540 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001246684.1 NP_001233613.1 3 - 4 949 97
GenBank mRNA AF113703.1 - 3 - 4 949 97

View Full Product Details