Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prss50
Entrez Gene ID
100771600
Gene Name
protease, serine, 50
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614270: 91343 - 97041 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007647210.1 XP_007645400.1 2 - 3 399 56

View Full Product Details