Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ms4a3
Entrez Gene ID
100765384
Gene Name
membrane-spanning 4-domains, subfamily A, member 3 (hematopoietic cell-specific)
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003616121: 107789 - 118700 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003513777.1 XP_003513825.1 1 - 2 159 120

View Full Product Details