Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Higd1b
Entrez Gene ID
100764860
Gene Name
HIG1 hypoxia inducible domain family, member 1B
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614071: 907577 - 910696 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003504529.2 XP_003504577.1 2 - 3 1530 116

View Full Product Details