Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Maff
Entrez Gene ID
100766059
Gene Name
v-maf avian musculoaponeurotic fibrosarcoma oncogene homolog F
Gene Aliases
I79_021907
Chromosome Location
Chr.Un|NW_003615689: 50899 - 61551 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003512999.2 XP_003513047.2 1 - 2 459 69

View Full Product Details