Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lactbl1
Entrez Gene ID
100770153
Gene Name
lactamase, beta-like 1
Gene Aliases
I79_010510
Chromosome Location
Chr.Un|NW_003613985: 1137682 - 1149619 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007644230.1 XP_007642420.1 1 - 2 146 106

View Full Product Details