Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ptpla
Entrez Gene ID
100772448
Gene Name
protein tyrosine phosphatase-like (proline instead of catalytic arginine), member A
Gene Aliases
I79_010168
Chromosome Location
Chr.Un|NW_003613962: 924555 - 936450 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003503378.2 XP_003503426.2 1 - 2 259 112
GenBank mRNA JP052801.1 - - 259 112

View Full Product Details