Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prss58
Entrez Gene ID
100751468
Gene Name
protease, serine, 58
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614308: 511381 - 515315 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003506953.1 XP_003507001.1 4 - 5 573 90

View Full Product Details