Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
B3gat2
Entrez Gene ID
100756696
Gene Name
beta-1,3-glucuronyltransferase 2 (glucuronosyltransferase S)
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613608: 3017362 - 3071729 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003495991.2 XP_003496039.1 1 - 2 100 73

View Full Product Details