Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Exoc8
Entrez Gene ID
100764733
Gene Name
exocyst complex component 8
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613636: 2196691 - 2200568 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003496809.2 XP_003496857.2 1 - 1 521 91

View Full Product Details