Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Aadacl3
Entrez Gene ID
100751803
Gene Name
arylacetamide deacetylase-like 3
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003616544: 84230 - 91612 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003514360.1 XP_003514408.1 1 - 2 169 70

View Full Product Details